3′-Sialyl-Lewis-X tetrasaccharide ≥95% (by TLC), Calbiochem®, Millipore®
Supplier: MilliporeSigma
Synonyms:
(2S,4S,5R,6R)-5-Acetamido-2-(((2S,3R,4S,5S,6R)-2-(((2R,3R,4R,5R)-5-acetamido-1,2-dihydroxy-6-oxo-4-(((2S,3S,4R,5S,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)hexan-3-yl)oxy)-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-4-yl)oxy)-4-hydroxy-6-((1R,2R)-1,2,3-trihydroxypropyl)tetrahydro-2H-pyran-2-carboxylic acid, Sialyl-Lewis X
565950-5MG
565950-1MG
80054-426EA
1136.75
USD
80054-426
80054-424
3′-Sialyl-Lewis-X tetrasaccharide ≥95% (by TLC), Calbiochem®, Millipore®
3′-Sialyl-Lewis-X tetrasaccharide
Sialyl Lewis x, Sodium Salt,Enzymatically prepared tetrasaccharide which acts as a minimal binding ligand for E-selectins.
Formula:
C₃₁H₅₂N₂O₂₃ MW: 820.74 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00270073 CAS Number: 98603-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...