NADP disodium salt ≥95% (by enzymatic purity), Calbiochem®, Millipore®

Supplier: MilliporeSigma

Synonyms: TPN, 2Na, β-Nicotinamide-adenine dinucleotide phosphate disodium salt

481972-1GM 481972-100MG
80108-202EA 524.48 USD
80108-202 80108-200
NADP disodium salt ≥95% (by enzymatic purity), Calbiochem®, Millipore®
NADP disodium salt

NADP, Disodium Salt, a coenzyme that is widely distributed in living matter. Participates in oxidation-reduction reactions.

Formula: C₂₁H₂₈N₇O₁₇P₃.2Na
MW: 789.39 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00065390
CAS Number: 24292-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR