Nystatin 95 HPLC_sum2peaks Streptomyces noursei, Calbiochem®, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Fungicidin, Mycostatin
475914-5GM
475914-1GM
80108-232EA
244.49
USD
80108-232
80057-066
Nystatin 95 HPLC_sum2peaks Streptomyces noursei, Calbiochem®, Sigma-Aldrich®
Nystatin
Nystatin, Streptomyces noursei, a polyene antifungal antibiotic effective against yeast and mycoplasma.
Formula:
C₄₇H₇₅NO₁₇ MW: 926.09 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD25541651 CAS Number: 1400-61-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...