L(+)-Cysteine hydrochloride ≥98% (by titration), Calbiochem®, Millipore®

Supplier: MilliporeSigma

Synonyms: H-Cys-OH.HCl, (R)-2-Amino-3-mercaptopropanoic acid hydrochloride, L-Cysteine hydrochloride, (R)-(+)-Cysteine hydrochloride

2430-100GM 2430-1KG
80057-974EA 138.41 USD
80057-974 80057-976
L(+)-Cysteine hydrochloride ≥98% (by titration), Calbiochem®, Millipore®
L(+)-Cysteine hydrochloride
Formula: HSCH₂CH(NH₂)COOH·HCl
Storage Temperature: Ambient
MDL Number: MFCD00064553
CAS Number: 52-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR