Nα-Benzyloxycarbonyl-L-lysine thiobenzyl ester hydrochloride ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Z-L-Lys-SBzl hydrochloride, BLT HCl, Z-Lys-SBzl·HCl, (S)-S-Benzyl 6-amino-2-(((benzyloxy)carbonyl)amino)hexanethioate hydrochloride

200274-100MG 200274-25MG
80055-860EA 243.92 USD
80055-860 80055-862
Nα-Benzyloxycarbonyl-L-lysine thiobenzyl ester hydrochloride ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Nα-Benzyloxycarbonyl-L-lysine thiobenzyl ester hydrochloride

Highly sensitive substrate for trypsin-like enzymes and granule-associated serine esterase.

Formula: C₂₁H₂₇ClN₂O₃S
MW: 422.968 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00058089
CAS Number: 69861-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR