Adenosine-5'-triphosphate disodium salt (ATP disodium salt) ≥95% (by HPLC), Calbiochem®, Millipore®
Supplier: MilliporeSigma
Synonyms:
((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt, Adenosine 5'-triphosphoric acid disodium, Adenosine 5′-triphosphate disodium salt, ATP 2Na, ATP disodium salt, Adenosine 5ʹ-Triphosphate, Disodium Salt
1191-25GM
1191-5GM
1191-50GM
1191-1GM
80050-120EA
435.78
USD
80050-120
80050-122
80108-478
80050-118
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) ≥95% (by HPLC), Calbiochem®, Millipore®
Adenosine 5'-triphosphate disodium salt (ATP disodium salt)
Formula:
C₁₀H₁₄N₅Na₂O₁₃P₃ MW: 551.14 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00080339 CAS Number: 987-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...