Accessories for Transport Case
Supplier: SARSTEDT INC
95.1714 95.1725 95.1712 95.1713
77589-962CS 143.34 USD
77589-962 77589-964 77589-958 77589-960
Accessories for Transport Case
Boxes Transport Boxes
The transport of diagnostics specimens of substance class UN 3373 is subject to legal regulations and guidelines. This range offers sample transport systems and secondary containers for a wide variety of transport requirements.
  • Superabsorber pack: High liquid absorption of approx. 1.2 L
  • Cross-bottom bag: Plastic bag with closure clip as secondary packaging
  • Closure clip: For optimal closure of the cross-bottom bag
  • Ice pack bag: Insulated bag for standard ice packs
For courier transport of diagnostic samples.