ASB-14 97 HPLC_ASSAY_METHOD Detergent_Zwitterionic, Calbiochem®, Millipore®

Supplier: MilliporeSigma

Synonyms: Amidosulphobetaine-14

MQ100

182750-5GM 182750-25GM
80501-334EA 296.27 USD
80501-334 80501-332
ASB-14 97 HPLC_ASSAY_METHOD Detergent_Zwitterionic, Calbiochem®, Millipore®
ASB-14

97

Formula: C₂₂H₄₆N₂O₄S
MW: 434.68 g/mol
Storage Temperature: Ambient
CAS Number: 216667-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR