Mailing Containers, Square
Supplier: SARSTEDT INC
78.572 78.573
77589-096CS 155.51 USD
77589-096 77589-098
Mailing Containers, Square
Mailers and Shippers
Sarstedt offers sample transport systems and secondary containers for a wide variety of transport requirements.
  • Corresponds to the requirement: P 650, ADR
  • Closure: Enclosed
  • Mailing container with screw cap made of HD-PE as a safe container wrapper for blood, urine, swab and stool sample tubes
  • Easy and quick opening and closing with a leak-proof screw cap
  • The square cross-sectional shape ensures optimal storage
  • Available with and without an absorbent liner
Mailing containers with screw cap; a secure container for blood, urine and stool sample tubes.