RecombiMAb Human IgG2 Isotype Control Anti-Hen Egg Lysozyme

Supplier: Bio X Cell
CP180-100MG CP180-1MG CP180-25MG CP180-50MG CP180-5MG
77580-460EA 6269.61 USD
77580-460 77580-462 77580-464 77580-466 77580-468
RecombiMAb Human IgG2 Isotype Control Anti-Hen Egg Lysozyme
Antibodies
Type: Control
Antigen: IgG2
Clonality: Recombinant
Clone: N/A-CP180
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: Hen
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR