RecombiMAb Human IgG1 (D265A) Isotype Control Anti-Hen Egg Lysozyme

Supplier: Bio X Cell
CP173-100MG CP173-1MG CP173-25MG CP173-50MG CP173-5MG
77580-420EA 6269.61 USD
77580-420 77580-422 77580-424 77580-426 77580-428
RecombiMAb Human IgG1 (D265A) Isotype Control Anti-Hen Egg Lysozyme
Antibodies
Type: Control
Antigen: IgG1
Clonality: Recombinant
Clone: N/A-CP173
Conjugation:
Epitope:
Host:
Isotype: IgG1
Reactivity: Hen
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR