RecombiMAb Anti-Ly6G/Ly6C Mouse Recombinant Antibody [clone: RB6-8C5-CP172]

Supplier: Bio X Cell
CP172-100MG CP172-1MG CP172-25MG CP172-50MG CP172-5MG
77580-410EA 6269.61 USD
77580-410 77580-412 77580-414 77580-416 77580-418
RecombiMAb Anti-Ly6G/Ly6C Mouse Recombinant Antibody [clone: RB6-8C5-CP172]
Antibodies
Type: Primary
Antigen: Ly6G/Ly6C
Clonality: Recombinant
Clone: RB6-8C5-CP172
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR