RecombiMAb mouse IgG2a (D265A) Isotype Control Anti-Hen Egg Lysozyme

Supplier: Bio X Cell
CP150-100MG CP150-1MG CP150-25MG CP150-50MG CP150-5MG
77580-250EA 6269.61 USD
77580-250 77580-252 77580-254 77580-256 77580-258
RecombiMAb mouse IgG2a (D265A) Isotype Control Anti-Hen Egg Lysozyme
Antibodies
Type: Control
Antigen: IgG2a
Clonality: Recombinant
Clone: N/A-CP150
Conjugation:
Epitope:
Host:
Isotype: IgG2a
Reactivity: Hen
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR