RecombiMAb mouse IgG2a (LALA-PG) Isotype Control Anti-Hen Egg Lysozyme

Supplier: Bio X Cell
CP048-100MG CP048-1MG CP048-25MG CP048-50MG CP048-5MG
77580-108EA 6269.61 USD
77580-108 77580-110 77580-112 77580-114 77580-116
RecombiMAb mouse IgG2a (LALA-PG) Isotype Control Anti-Hen Egg Lysozyme
Antibodies
Type: Control
Antigen: IgG2a
Clonality: Recombinant
Clone: N/A-CP048
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: Hen
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR