RecombiMAb mouse IgG1 (D265A) Isotype Control Anti-Hen Egg Lysozyme

Supplier: Bio X Cell
CP047-100MG CP047-1MG CP047-25MG CP047-50MG CP047-5MG
77580-098EA 6269.61 USD
77580-098 77580-100 77580-102 77580-104 77580-106
RecombiMAb mouse IgG1 (D265A) Isotype Control Anti-Hen Egg Lysozyme
Antibodies
Type: Control
Antigen: IgG1
Clonality: Recombinant
Clone: N/A-CP047
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: Hen
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR