InVivoMAb Anti-S protein (RBD) Mouse Monoclonal Antibody [clone: SARS2-38]

Supplier: Bio X Cell
BE0438-100MG BE0438-1MG BE0438-25MG BE0438-50MG BE0438-5MG
77579-920EA 4408.4 USD
77579-920 77579-922 77579-924 77579-926 77579-928
InVivoMAb Anti-S protein (RBD) Mouse Monoclonal Antibody [clone: SARS2-38]
Antibodies
Type: Primary
Antigen: S protein (RBD)
Clonality: Monoclonal
Clone: SARS2-38
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: SARS-CoV-2
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR