InVivoMAb Anti-33D1 Rat Monoclonal Antibody [clone: 33D1]

Supplier: Bio X Cell
BE0426-100MG BE0426-1MG BE0426-25MG BE0426-50MG BE0426-5MG
77579-800EA 4588.65 USD
77579-800 77579-802 77579-804 77579-806 77579-808
InVivoMAb Anti-33D1 Rat Monoclonal Antibody [clone: 33D1]
Antibodies
Type: Primary
Antigen: 33D1
Clonality: Monoclonal
Clone: 33D1
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR