InVivoMAb Anti-Mesothelin Rabbit Monoclonal Antibody [clone: YP218]

Supplier: Bio X Cell
BE0405-100MG BE0405-1MG BE0405-25MG BE0405-50MG BE0405-5MG
77579-590EA 4408.4 USD
77579-590 77579-592 77579-594 77579-596 77579-598
InVivoMAb Anti-Mesothelin Rabbit Monoclonal Antibody [clone: YP218]
Antibodies
Type: Primary
Antigen: Mesothelin
Clonality: Monoclonal
Clone: YP218
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR