InVivoMAb Anti-GRP78 Mouse Monoclonal Antibody [clone: C107]

Supplier: Bio X Cell
BE0397-100MG BE0397-1MG BE0397-25MG BE0397-50MG BE0397-5MG
77579-510EA 4408.4 USD
77579-510 77579-512 77579-514 77579-516 77579-518
InVivoMAb Anti-GRP78 Mouse Monoclonal Antibody [clone: C107]
Antibodies
Type: Primary
Antigen: GRP78
Clonality: Monoclonal
Clone: C107
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR