InVivoMAb Anti-FAP Mouse Monoclonal Antibody [clone: 73.3]

Supplier: Bio X Cell
BE0374-100MG BE0374-1MG BE0374-25MG BE0374-50MG BE0374-5MG
77579-280EA 4408.4 USD
77579-280 77579-282 77579-284 77579-286 77579-288
InVivoMAb Anti-FAP Mouse Monoclonal Antibody [clone: 73.3]
Antibodies
Type: Primary
Antigen: FAP
Clonality: Monoclonal
Clone: 73.3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR