RecombiMAb Human IgG4 (S228P) Isotype control Anti-Hen Egg Lysozyme

Supplier: Bio X Cell
CP181-100MG CP181-1MG CP181-25MG CP181-50MG CP181-5MG
77580-470EA 6269.61 USD
77580-470 77580-472 77580-474 77580-476 77580-478
RecombiMAb Human IgG4 (S228P) Isotype control Anti-Hen Egg Lysozyme
Antibodies
Type: Control
Antigen: IgG4
Clonality: Recombinant
Clone: N/A-CP181
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: Hen
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR