RecombiMAb Human IgG1 (K214R/L234F/L235E/P331S) Isotype control Anti-Hen Egg Lysozyme

Supplier: Bio X Cell
CP175-100MG CP175-1MG CP175-25MG CP175-50MG CP175-5MG
77580-440EA 6269.61 USD
77580-440 77580-442 77580-444 77580-446 77580-448
RecombiMAb Human IgG1 (K214R/L234F/L235E/P331S) Isotype control Anti-Hen Egg Lysozyme
Antibodies
Type: Control
Antigen: IgG1
Clonality: Recombinant
Clone: N/A-CP175
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: Hen
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR