RecombiMAb Human IgG1 Isotype control Anti-Hen Egg Lysozyme

Supplier: Bio X Cell
CP174-100MG CP174-1MG CP174-25MG CP174-50MG CP174-5MG
77580-430EA 6269.61 USD
77580-430 77580-432 77580-434 77580-436 77580-438
RecombiMAb Human IgG1 Isotype control Anti-Hen Egg Lysozyme
Antibodies
Type: Control
Antigen: IgG1
Clonality: Recombinant
Clone: N/A-CP174
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: Hen
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR