RecombiMAb Human IgG1 Isotype control Anti-Respiratory Syncytial Virus

Supplier: Bio X Cell
CP169-100MG CP169-1MG CP169-25MG CP169-50MG CP169-5MG
77580-390EA 6269.61 USD
77580-390 77580-392 77580-394 77580-396 77580-398
RecombiMAb Human IgG1 Isotype control Anti-Respiratory Syncytial Virus
Antibodies
Type: Control
Antigen: IgG1
Clonality: Recombinant
Clone: Palivizumab-CP169
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR