InVivoSIM Anti-IL-4Ra Human Antibody

Supplier: Bio X Cell
SIM0024-100MG SIM0024-1MG SIM0024-25MG SIM0024-50MG SIM0024-5MG
77580-740EA 7984.56 USD
77580-740 77580-742 77580-744 77580-746 77580-748
InVivoSIM Anti-IL-4Ra Human Antibody
Antibodies
Vedolizumab biosimilar.

Type:
Antigen: IL-4Ra
Clonality:
Clone:
Conjugation:
Epitope:
Host: Human
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR