InVivoSIM Anti-CTLA-4 Human Antibody

Supplier: Bio X Cell
SIM0004-100MG SIM0004-1MG SIM0004-25MG SIM0004-50MG SIM0004-5MG
77580-540EA 7984.56 USD
77580-540 77580-542 77580-544 77580-546 77580-548
InVivoSIM Anti-CTLA-4 Human Antibody
Antibodies
Ipilimumab biosimilar.

Type:
Antigen: CTLA-4
Clonality:
Clone:
Conjugation:
Epitope:
Host: Human
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR