RecombiMAb Anti-CD8a Mouse Recombinant Antibody [clone: YTS]

Supplier: Bio X Cell
CP134-100MG CP134-1MG CP134-25MG CP134-50MG CP134-5MG
77580-190EA 6269.61 USD
77580-190 77580-192 77580-194 77580-196 77580-198
RecombiMAb Anti-CD8a Mouse Recombinant Antibody [clone: YTS]
Antibodies
Type: Primary
Antigen: CD8a
Clonality: Recombinant
Clone: YTS
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR