InVivoPlus Anti-CD4 Mouse Antibody [clone: OX-38]

Supplier: Bio X Cell
BP0308-100MG BP0308-25MG BP0308-50MG BP0308-5MG
77580-008EA 5735.04 USD
77580-008 77580-010 77580-012 77580-014
InVivoPlus Anti-CD4 Mouse Antibody [clone: OX-38]
Antibodies
Type: Primary
Antigen: CD4
Clonality:
Clone: OX-38
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR