InVivoMAb Anti-CD4 Mouse Monoclonal Antibody [clone: W3/25]

Supplier: Bio X Cell
BE0439-100MG BE0439-1MG BE0439-25MG BE0439-50MG BE0439-5MG
77579-930EA 4408.4 USD
77579-930 77579-932 77579-934 77579-936 77579-938
InVivoMAb Anti-CD4 Mouse Monoclonal Antibody [clone: W3/25]
Antibodies
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: W3/25
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR