InVivoMAb Anti-myeloperoxidase Mouse Monoclonal Antibody [clone: 6G4]

Supplier: Bio X Cell
BE0392-100MG BE0392-1MG BE0392-25MG BE0392-50MG BE0392-5MG
77579-460EA 4438 USD
77579-460 77579-462 77579-464 77579-466 77579-468
InVivoMAb Anti-myeloperoxidase Mouse Monoclonal Antibody [clone: 6G4]
Antibodies
Type: Primary
Antigen: myeloperoxidase
Clonality: Monoclonal
Clone: 6G4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR