InVivoMAb Anti-CD71 (TfR1) Mouse Monoclonal Antibody [clone: BK19.9]

Supplier: Bio X Cell
BE0388-100MG BE0388-1MG BE0388-25MG BE0388-50MG BE0388-5MG
77579-420EA 4408.4 USD
77579-420 77579-422 77579-424 77579-426 77579-428
InVivoMAb Anti-CD71 (TfR1) Mouse Monoclonal Antibody [clone: BK19.9]
Antibodies
Type: Primary
Antigen: CD71 (TfR1)
Clonality: Monoclonal
Clone: BK19.9
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR