InVivoMAb Anti-CD326 Rat Monoclonal Antibody [clone: G8.8]

Supplier: Bio X Cell
BE0346-100MG BE0346-1MG BE0346-25MG BE0346-50MG BE0346-5MG
77579-010EA 4408.4 USD
77579-010 77579-012 77579-014 77579-016 77579-018
InVivoMAb Anti-CD326 Rat Monoclonal Antibody [clone: G8.8]
Antibodies
Type: Primary
Antigen: CD326
Clonality: Monoclonal
Clone: G8.8
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR