2-(3-(Thiophene-2-carbonyl)phenyl)propanoic acid

Supplier: TCI America

Synonyms: Suprofen, 2-(4-(Thiophene-2-carbonyl)phenyl)propanoic acid

U0138-100MG U0138-25MG
TCU0138-100MGEA 370.8 USD
TCU0138-100MG TCU0138-25MG
2-(3-(Thiophene-2-carbonyl)phenyl)propanoic acid
2-(3-(Thiophene-2-carbonyl)phenyl)propanoic acid
Formula: C₁₄H₁₂O₃S
MW: 260.30 g/mol
MDL Number: MFCD00079572
CAS Number: 40828-46-4
UN: 2811

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR