(3R,4R,5R)-3,4-Bis(benzyloxy)-5-((benzyloxy)methyl)dihydrofuran-2(3H)-one

Supplier: TCI America

Synonyms: 2,3,5-Tri-O-benzyl-D-ribono-1,4-lactone

T3955-25G T3955-5G
TCT3955-25GEA 297.67 USD
TCT3955-25G TCT3955-5G
(3R,4R,5R)-3,4-Bis(benzyloxy)-5-((benzyloxy)methyl)dihydrofuran-2(3H)-one
(3R,4R,5R)-3,4-Bis(benzyloxy)-5-((benzyloxy)methyl)dihydrofuran-2(3H)-one
Formula: C₂₆H₂₆O₅
MW: 418.48 g/mol
MDL Number: MFCD08703966
CAS Number: 55094-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR