2,2,6-Trimethylcyclohexanone

Supplier: TCI America

T3944-1G T3944-5G
TCT3944-1GEA 82.4 USD
TCT3944-1G TCT3944-5G
2,2,6-Trimethylcyclohexanone
2,2,6-Trimethylcyclohexanone
Formula: C₉H₁₆O
MDL Number: MFCD00045550
CAS Number: 2408-37-9
UN: 1224

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR