[6-(Trifluoromethyl)-3-pyridinyl]methanol

Supplier: TCI America

Synonyms: 5-(Hydroxymethyl)-2-(trifluoromethyl)pyridine, [6-(Trifluoromethyl)pyridin-3-yl]methanol

T3927-1G T3927-250MG
TCT3927-1GEA 114.33 USD
TCT3927-1G TCT3927-250MG
[6-(Trifluoromethyl)-3-pyridinyl]methanol
[6-(Trifluoromethyl)-3-pyridinyl]methanol
Formula: C₇H₆F₃NO
MW: 177.13 g/mol
MDL Number: MFCD01862648
CAS Number: 386704-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR