1,3-Thiazolan-2-one

Supplier: TCI America

T3926-25G T3926-5G
TCT3926-25GEA 127.72 USD
TCT3926-25G TCT3926-5G
1,3-Thiazolan-2-one
1,3-Thiazolan-2-one
Formula: C₃H₅NOS
MDL Number: MFCD01332388
CAS Number: 2682-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR