4-(2H-Tetrazol-5-yl)benzoic acid

Supplier: TCI America

Synonyms: 4-(1H-Tetrazol-5-yl)benzoic acid

T3917-1G T3917-200MG
TCT3917-1GEA 387.28 USD
TCT3917-1G TCT3917-200MG
4-(2H-Tetrazol-5-yl)benzoic acid
4-(2H-Tetrazol-5-yl)benzoic acid
Formula: C₈H₆N₄O₂
MW: 190.16 g/mol
MDL Number: MFCD01569290
CAS Number: 34114-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR