Octaethyl (benzene-1,2,4,5-tetrayltetrakis(methylene))tetrakis(phosphonate)

Supplier: TCI America

Synonyms: Octaethyl [benzene-1,2,4,5-tetrayltetrakis(methylene)]tetrakis (phosphonate)

T3904-1G T3904-5G
TCT3904-1GEA 64.89 USD
TCT3904-1G TCT3904-5G
Octaethyl (benzene-1,2,4,5-tetrayltetrakis(methylene))tetrakis(phosphonate)
Octaethyl (benzene-1,2,4,5-tetrayltetrakis(methylene))tetrakis(phosphonate)
Formula: C₂₆H₅₀O₁₂P₄
MW: 678.56 g/mol
CAS Number: 136455-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR