1,2,4,5-Tetrakis(bromomethyl)benzene

Supplier: TCI America

T3901-1G T3901-5G
TCT3901-1GEA 69.01 USD
TCT3901-1G TCT3901-5G
1,2,4,5-Tetrakis(bromomethyl)benzene
1,2,4,5-Tetrakis(bromomethyl)benzene
Formula: C₁₀H₁₀Br₄
MW: 449.8 g/mol
MDL Number: MFCD00009664
CAS Number: 15442-91-8
UN: 3261

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR