4-[(Trimethylsilyl)ethynyl]benzaldehyde

Supplier: TCI America

Synonyms: 4-((Trimethylsilyl)ethynyl)benzaldehyde, 4-(Trimethylsilylethynyl)benzaldehyde

T3827-1G T3827-5G
TCT3827-1GEA 50.47 USD
TCT3827-1G TCT3827-5G
4-[(Trimethylsilyl)ethynyl]benzaldehyde
4-[(Trimethylsilyl)ethynyl]benzaldehyde
Formula: C₁₂H₁₄OSi
MDL Number: MFCD02093765
CAS Number: 77123-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR