Triamcinolone diacetate

Supplier: TCI America

T3676-250MG T3676-50MG
TCT3676-250MGEA 225.57 USD
TCT3676-250MG TCT3676-50MG
Triamcinolone diacetate
Triamcinolone diacetate
Formula: C₂₅H₃₁FO₈
MDL Number: MFCD00056462
CAS Number: 67-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR