trans-4'-(p-Tolyl)-[1,1'-bi(cyclohexan)]-4-one ≥98.0% (by GC)

Supplier: TCI America

Synonyms: trans-4'-(4-Methylphenyl)[1,1'-bi(cyclohexyl)]-4-one, (1'r,4'r)-4'-(p-Tolyl)-[1,1'-bi(cyclohexan)]-4-one, trans-4'-(p-tolyl)-[1,1'-Bi(cyclohexan)]-4-one

T3465-1G T3465-5G
TCT3465-1GEA 47.38 USD
TCT3465-1G TCT3465-5G
trans-4'-(p-Tolyl)-[1,1'-bi(cyclohexan)]-4-one ≥98.0% (by GC)
trans-4'-(p-Tolyl)-[1,1'-bi(cyclohexan)]-4-one
Formula: C₁₉H₂₆O
MW: 270.42 g/mol
MDL Number: MFCD13194928
CAS Number: 125962-80-3
EINECS: 435-110-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 121.0 to 125.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR