4,4',4''-Tris[9,9-dimethylfluoren-2-yl(phenyl)amino]triphenylamine ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: N-(9,9-Dimethyl-9H-fluoren-2-yl)-N',N'-bis[4-[(9,9-dimethyl-9H-fluoren-2-yl)(phenyl)amino]phenyl]-N-phenyl-1,4-benzenediamine

T3438-1G T3438-200MG
TCT3438-1GEA 470.71 USD
TCT3438-1G TCT3438-200MG
4,4',4''-Tris[9,9-dimethylfluoren-2-yl(phenyl)amino]triphenylamine ≥98.0% (by HPLC)
4,4',4''-Tris[9,9-dimethylfluoren-2-yl(phenyl)amino]triphenylamine
Formula: C₈₁H₆₆N₄
MW: 1095.45 g/mol
CAS Number: 303111-06-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR