4-(p-Tolyloxy)benzoic acid

Supplier: TCI America

Synonyms: 4-(4-Methylphenoxy)benzoic acid

T3391-1G T3391-5G
TCT3391-1GEA 128.75 USD
TCT3391-1G TCT3391-5G
4-(p-Tolyloxy)benzoic acid
4-(p-Tolyloxy)benzoic acid
Formula: C₁₄H₁₂O₃
MW: 228.24 g/mol
MDL Number: MFCD01631893
CAS Number: 21120-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR