N-Methyl-3-(triethoxysilyl)propylamine

Supplier: TCI America

Synonyms: N-Methyl-3-(triethoxysilyl)propan-1-amine, [3-(Methylamino)prop-1-yl]triethoxysilane

T3242-25G T3242-5G
TCT3242-25GEA 978.5 USD
TCT3242-25G TCT3242-5G
N-Methyl-3-(triethoxysilyl)propylamine
N-Methyl-3-(triethoxysilyl)propylamine
Formula: C₁₀H₂₅NO₃Si
MW: 235.396 g/mol
MDL Number: MFCD24842713
CAS Number: 6044-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR