2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (2-Pyrenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-Tetramethyl-2-(pyren-2-yl)-1,3,2-dioxaborolane, 2-Pyreneboronic acid Pinacol ester

T3150-1G T3150-5G
TCT3150-1GEA 404.79 USD
TCT3150-1G TCT3150-5G
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene ≥98.0% (by GC)
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene
Formula: C₂₂H₂₁BO₂
MW: 328.212 g/mol
MDL Number: MFCD11977261
CAS Number: 853377-11-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 126.0 to 130.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR