Tetrahydrozoline hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-(1,2,3,4-Tetrahydronaphthalen-1-yl)-4,5-dihydro-1H-imidazole hydrochloride

T3148-25G T3148-5G
TCT3148-25GEA 590.19 USD
TCT3148-25G TCT3148-5G
Tetrahydrozoline hydrochloride ≥98.0% (by HPLC, titration analysis)
Tetrahydrozoline hydrochloride
Formula: C₁₃H₁₇ClN₂
MW: 236.7405 g/mol
MDL Number: MFCD00058029
CAS Number: 522-48-5
EINECS: 208-329-3
Merck Index: 14,09219

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR