Sodium 3,5,6-trichloropyridin-2-olate
Supplier: TCI America
T3004-1G
T3004-5G
TCT3004-1GEA
43.26
USD
TCT3004-1G
TCT3004-5G
Sodium 3,5,6-trichloropyridin-2-olate
Sodium 3,5,6-trichloropyridin-2-olate
Formula:
C₅HCl₃NNaO |
MDL Number:
MFCD08704561 CAS Number: 37439-34-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...