(S)-5-Benzyl-2,2,3-trimethylimidazolidin-4-one hydrochloride

Supplier: TCI America

Synonyms: (S)-(-)-5-Benzyl-2,2,3-trimethyl-4-imidazolidinone hydrochloride

T2991-1G T2991-250MG
TCT2991-1GEA 181.28 USD
TCT2991-1G TCT2991-250MG
(S)-5-Benzyl-2,2,3-trimethylimidazolidin-4-one hydrochloride
(S)-5-Benzyl-2,2,3-trimethylimidazolidin-4-one hydrochloride
Formula: C₁₃H₁₉ClN₂O
MW: 254.76 g/mol
MDL Number: MFCD03426983
CAS Number: 278173-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR